siRNA Details
| Gene Symbol: TMEM140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018295.4 | NP_060765.4 | 2 | 1022 | ||
| GenBank mRNA | AK001862.1 | - | 2 | 964 | ||
| AK225055.1 | - | 2 | 952 | |||
| AK301268.1 | - | 3 | 838 | |||
| BC020942.1 | - | 2 | 949 | 4798940 | ||
| BM512090.1 | - | 1 | 279 | |||

TMEM140
Genomic Map