siRNA Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018342.4 | NP_060812.2 | 10, 11 | 1318 | ||
| XM_005263110.1 | XP_005263167.1 | 11, 12 | 1273 | |||
| XM_005263112.3 | XP_005263169.1 | 12, 13 | 1308 | |||
| XM_006714254.1 | XP_006714317.1 | 11, 12 | 1130 | |||
| XM_017008366.1 | XP_016863855.1 | 9, 10 | 1286 | |||
| XM_017008367.1 | XP_016863856.1 | 8, 9 | 858 | |||
| XM_017008368.1 | XP_016863857.1 | 9, 10 | 1018 | |||
| GenBank mRNA | AK002017.1 | - | 9, 10 | 1222 | ||
| BC026318.1 | - | 9, 10 | 944 | |||
| BC054487.1 | - | 10, 11 | 1066 | 5732836 | ||
| BC060875.1 | - | 10, 11 | 1044 | 4800219 | ||

TMEM144
Genomic Map