siRNA Details
| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018392.4 | NP_060862.3 | 5 | 388 | ||
| XM_005263115.3 | XP_005263172.1 | 5 | 443 | |||
| XM_011532091.2 | XP_011530393.1 | 5 | 388 | |||
| XM_011532092.2 | XP_011530394.1 | 4 | 268 | |||
| XM_011532093.2 | XP_011530395.1 | 5 | 388 | |||
| XM_011532094.2 | XP_011530396.1 | 5 | 388 | |||
| XM_011532096.2 | XP_011530398.1 | 5 | 388 | |||
| XM_017008369.1 | XP_016863858.1 | 4 | 383 | |||
| XM_017008370.1 | XP_016863859.1 | 5 | 388 | |||
| XM_017008371.1 | XP_016863860.1 | 4 | 268 | |||
| XM_017008372.1 | XP_016863861.1 | 5 | 297 | |||
| XM_017008373.1 | XP_016863862.1 | 4 | 234 | |||
| GenBank mRNA | AK002193.1 | - | 5 | 305 | ||
| AK299831.1 | - | 5 | 388 | |||
| AK302613.1 | - | 6 | 613 | |||
| AL708767.1 | - | 5 | 311 | |||
| BC044799.1 | - | 5 | 293 | 4836807 | ||

ZGRF1
Genomic Map