siRNA Details
| Gene Symbol: TMEM176A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018487.2 | NP_060957.2 | 6 | 655 | ||
| XM_011516376.2 | XP_011514678.1 | 6 | 790 | |||
| XM_011516377.2 | XP_011514679.1 | 6 | 793 | |||
| XM_017012393.1 | XP_016867882.1 | 6 | 878 | |||
| GenBank mRNA | AF258340.1 | - | 6 | 630 | ||
| BC008303.1 | - | 6 | 673 | 3536986 | ||

TMEM176A
Genomic Map