siRNA Details
| Gene Symbol: SAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018666.2 | NP_061136.2 | 2, 3 | 262 | ||
| XM_017029621.1 | XP_016885110.1 | 2, 3 | 270 | |||
| XM_017029622.1 | XP_016885111.1 | 2, 3 | 181 | |||
| XM_017029623.1 | XP_016885112.1 | 2, 3 | 177 | |||
| XM_017029624.1 | XP_016885113.1 | 2, 3 | 177 | |||
| XM_017029625.1 | XP_016885114.1 | 2, 3 | 177 | |||
| GenBank mRNA | AJ278111.1 | - | 2, 3 | 262 | ||
| AK310412.1 | - | 2, 3 | 311 | |||
| BC144260.1 | - | 2, 3 | 143 | 9052780 | ||

SAGE1
Genomic Map