siRNA Details
| Gene Symbol: ZC4H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178032.2 | NP_001171503.1 | 3, 4 | 713 | ||
| NM_001243804.1 | NP_001230733.1 | 3, 4 | 458 | |||
| NM_018684.3 | NP_061154.1 | 3, 4 | 551 | |||
| NR_045044.1 | 3, 4 | 806 | ||||
| GenBank mRNA | AF270491.1 | - | 3, 4 | 483 | ||
| AK022807.1 | - | 3, 4 | 682 | |||
| AK022918.1 | - | 3, 4 | 445 | |||
| AK123763.1 | - | 4, 5 | 944 | |||
| AK222708.1 | - | 3, 4 | 468 | |||
| AK315485.1 | - | 3, 4 | 466 | |||
| BC004411.1 | - | 3, 4 | 502 | 3639978 | ||
| BI770655.1 | - | 3, 4 | 636 | |||
| BP376634.1 | - | 3, 4 | 458 | |||
| CX782941.1 | - | 3, 4 | 423 | |||

ZC4H2
Genomic Map