siRNA Details
| Gene Symbol: PUS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318163.1 | NP_001305092.1 | 16 | 2048 | ||
| NM_001318164.1 | NP_001305093.1 | 16 | 2237 | |||
| NM_019042.4 | NP_061915.2 | 16 | 2261 | |||
| XM_005250462.3 | XP_005250519.1 | 17 | 2287 | |||
| XM_011516336.2 | XP_011514638.1 | 16 | 2180 | |||
| XM_017012367.1 | XP_016867856.1 | 17 | 2215 | |||
| XM_017012368.1 | XP_016867857.1 | 14 | 2026 | |||
| GenBank mRNA | AB067484.1 | - | 16 | 1975 | ||
| AK000492.1 | - | 6 | 765 | |||
| AK128629.1 | - | 16 | 2223 | |||
| BC005209.2 | - | 8 | 974 | |||
| BC011396.1 | - | 16 | 2217 | 3870711 | ||

PUS7
Genomic Map