siRNA Details
| Gene Symbol: INTS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142471.1 | NP_001135943.1 | 3 | 314 | ||
| NM_020395.3 | NP_065128.2 | 4 | 476 | |||
| XM_005263148.4 | XP_005263205.1 | 3 | 969 | |||
| XM_011532143.2 | XP_011530445.1 | 4 | 1125 | |||
| XM_011532145.2 | XP_011530447.1 | 5 | 612 | |||
| GenBank mRNA | AF242524.1 | - | 3 | 308 | ||
| AK314939.1 | - | 3 | 314 | |||
| BC014442.1 | - | 4 | 424 | 4866321 | ||

INTS12
Genomic Map