siRNA Details
| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020961.3 | NP_066012.1 | 2 | 262 | ||
| XM_005263166.4 | XP_005263223.1 | 2 | 257 | |||
| XM_011532162.2 | XP_011530464.1 | 2 | 257 | |||
| GenBank mRNA | AB046847.1 | - | 2 | 217 | ||
| AK055555.1 | - | 2 | 240 | |||
| BC006565.2 | - | 2 | 213 | 3010654 | ||
| BC007449.2 | - | 2 | 213 | 3010654 | ||
| BG773151.1 | - | 2 | 267 | |||

METTL14
Genomic Map