siRNA Details
| Gene Symbol: CCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021174.5 | NP_066997.3 | 14 | 2088 | ||
| NR_033902.1 | 14 | 1906 | ||||
| XM_011544603.1 | XP_011542905.1 | 14 | 1906 | |||
| XM_011544604.1 | XP_011542906.1 | 13 | 1786 | |||
| XM_017013717.1 | XP_016869206.1 | 14 | 2088 | |||
| GenBank mRNA | AB075847.1 | - | 11 | 1397 | ||
| AF293335.1 | - | 3 | 344 | |||
| AK095712.1 | - | 14 | 1762 | |||
| AK096547.1 | - | 5 | 737 | |||
| AK314535.1 | - | 14 | 1781 | |||
| AL834162.1 | - | 14 | 2075 | |||
| AL834351.1 | - | 14 | 2076 | |||
| AL834352.1 | - | 14 | 2076 | |||
| BC018269.1 | - | 2 | 110 | |||
| BC065495.1 | - | 14 | 1764 | 5496068 | ||
| BX640952.1 | - | 14 | 1775 | |||

CCAR2
Genomic Map