siRNA Details
| Gene Symbol: TMEM35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021637.2 | NP_067650.1 | 2 | 1675 | ||
| GenBank mRNA | AK024146.1 | - | 2 | 1646 | ||
| AK223240.1 | - | 2 | 1642 | |||
| BC078658.1 | - | 2 | 1640 | 5261160 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM35A| Gene Symbol: TMEM35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021637.2 | NP_067650.1 | 2 | 1675 | ||
| GenBank mRNA | AK024146.1 | - | 2 | 1646 | ||
| AK223240.1 | - | 2 | 1642 | |||
| BC078658.1 | - | 2 | 1640 | 5261160 | ||