siRNA Details
| Gene Symbol: CXorf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170569.1 | NP_001164040.1 | 2, 3 | 413 | ||
| NM_022101.3 | NP_071384.1 | 2, 3 | 331 | |||
| GenBank mRNA | AK096297.1 | - | 2, 3 | 377 | ||
| BC023506.2 | - | 2, 3 | 292 | |||
| BQ223940.1 | - | 2, 3 | 296 | |||
| DA590933.1 | - | 2, 3 | 415 | |||
| DA921725.1 | - | 2, 3 | 306 | |||
| DB033119.1 | - | 2, 3 | 346 | |||

CXorf56
Genomic Map