siRNA Details
| Gene Symbol: CXorf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170569.1 | NP_001164040.1 | 7 | 873 | ||
| NM_001170570.1 | NP_001164041.1 | 6 | 749 | |||
| NM_022101.3 | NP_071384.1 | 7 | 791 | |||
| GenBank mRNA | AK096297.1 | - | 7 | 837 | ||
| AK298877.1 | - | 6 | 727 | |||
| BC023506.2 | - | 7 | 752 | |||
| BQ223940.1 | - | 7 | 756 | |||

CXorf56
Genomic Map