siRNA Details
| Gene Symbol: TSPYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022117.3 | NP_071400.1 | 7 | 2796 | ||
| XM_017029727.1 | XP_016885216.1 | 6 | 2961 | |||
| GenBank mRNA | AB209182.1 | - | 2 | 4153 | ||
| AF254794.1 | - | 7 | 2530 | |||
| AF273046.1 | - | 7 | 2786 | |||
| AK292462.1 | - | 7 | 2789 | |||
| AL713652.1 | - | 7 | 2775 | |||
| AY040871.1 | - | 7 | 2785 | |||
| BC001566.1 | - | 4 | 1593 | |||
| BC024270.1 | - | 7 | 2776 | 2819684 | ||

TSPYL2
Genomic Map