siRNA Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022483.4 | NP_071928.2 | 3 | 2030 | ||
| XM_005248333.4 | XP_005248390.1 | 4 | 2898 | |||
| XM_005248335.3 | XP_005248392.1 | 4 | 2182 | |||
| XM_006714486.3 | XP_006714549.1 | 3 | 2502 | |||
| XM_011514074.2 | XP_011512376.1 | 5 | 3274 | |||
| XM_011514075.2 | XP_011512377.1 | 5 | 3270 | |||
| GenBank mRNA | AL137306.1 | - | 1 | 1285 | ||
| BC013351.2 | - | 4 | 2219 | 3621124 | ||
| BC063832.1 | - | 2 | 1976 | 6178186 | ||
| BC073852.1 | - | 2 | 1963 | |||

TMEM267
Genomic Map