siRNA Details
| Gene Symbol: CCDC136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022742.4 | NP_073579.4 | 13 | 2736 | ||
| XM_005250534.3 | XP_005250591.1 | 14 | 2520 | |||
| XM_005250538.1 | XP_005250595.1 | 14 | 2520 | |||
| XM_011516483.1 | XP_011514785.1 | 15 | 2592 | |||
| XM_011516484.1 | XP_011514786.1 | 15 | 2592 | |||
| XM_011516485.1 | XP_011514787.1 | 15 | 2589 | |||
| XM_011516486.1 | XP_011514788.1 | 15 | 2556 | |||
| XM_011516487.2 | XP_011514789.1 | 15 | 2592 | |||
| XM_011516488.2 | XP_011514790.1 | 15 | 2592 | |||
| XM_011516489.1 | XP_011514791.1 | 15 | 2592 | |||
| XM_011516490.2 | XP_011514792.1 | 15 | 2611 | |||
| XM_017012532.1 | XP_016868021.1 | 14 | 2484 | |||
| XM_017012533.1 | XP_016868022.1 | 14 | 2986 | |||
| XM_017012534.1 | XP_016868023.1 | 14 | 2484 | |||
| GenBank mRNA | AB058696.1 | - | 13 | 2687 | ||
| AK023500.1 | - | 6 | 1093 | |||
| AL133027.1 | - | 10 | 1950 | |||
| BC070114.1 | - | 10 | 2473 | |||
| BC150331.1 | - | 13 | 2698 | 8860498 | ||
| BC152408.1 | - | 13 | 2687 | 8862511 | ||

CCDC136
Genomic Map