siRNA Details
| Gene Symbol: GPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127235.2 | NP_001120707.2 | 7 | 1667 | ||
| NM_001127236.2 | NP_001120708.1 | 6 | 714 | |||
| NM_001203246.1 | NP_001190175.1 | 6 | 1790 | |||
| NM_022913.3 | NP_075064.1 | 7 | 1910 | |||
| XM_006714673.3 | XP_006714736.1 | 9 | 2046 | |||
| XM_006714674.3 | XP_006714737.1 | 8 | 2025 | |||
| XM_017009754.1 | XP_016865243.1 | 8 | 1399 | |||
| XM_017009755.1 | XP_016865244.1 | 7 | 1378 | |||
| XM_017009756.1 | XP_016865245.1 | 7 | 1339 | |||
| XM_017009757.1 | XP_016865246.1 | 8 | 1986 | |||
| XM_017009758.1 | XP_016865247.1 | 7 | 1965 | |||
| XM_017009759.1 | XP_016865248.1 | 7 | 1762 | |||
| GenBank mRNA | AK024807.1 | - | 1 | 149 | ||
| AK307505.1 | - | 5 | 563 | |||
| AL136844.1 | - | 6 | 667 | |||
| AL161991.1 | - | 6 | 1689 | |||
| AY226828.1 | - | 7 | 1741 | |||
| BC000267.1 | - | 7 | 1169 | |||
| BC070132.1 | - | 3 | 393 | |||
| BC113004.2 | - | 6 | 1659 | 40034860 | ||
| CD657101.1 | - | 5 | 559 | |||

GPBP1
Genomic Map