siRNA Details
| Gene Symbol: FASTKD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024091.3 | NP_076996.2 | 2 | 1050 | ||
| XM_006714498.1 | XP_006714561.1 | 2 | 961 | |||
| GenBank mRNA | AK026927.1 | - | 2 | 1015 | ||
| BC001295.1 | - | 1 | 542 | |||
| BC113563.1 | - | 1 | 965 | 8322615 | ||
| BC143517.1 | - | 1 | 965 | |||
| BC143518.1 | - | 2 | 402 | |||

FASTKD3
Genomic Map