siRNA Details
| Gene Symbol: PARP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178055.1 | NP_001171526.1 | 10 | 809 | ||
| NM_001178056.1 | NP_001171527.1 | 9 | 785 | |||
| NM_024615.3 | NP_078891.2 | 9 | 785 | |||
| XM_005248596.4 | XP_005248653.1 | 10 | 1165 | |||
| XM_011543631.2 | XP_011541933.1 | 10 | 698 | |||
| XM_011543632.2 | XP_011541934.2 | 10 | 745 | |||
| XM_011543633.2 | XP_011541935.2 | 10 | 745 | |||
| XM_011543634.2 | XP_011541936.1 | 11 | 1294 | |||
| XM_011543635.2 | XP_011541937.1 | 11 | 815 | |||
| XM_011543636.2 | XP_011541938.1 | 10 | 679 | |||
| XM_017009850.1 | XP_016865339.1 | 9 | 1245 | |||
| XM_017009851.1 | XP_016865340.1 | 8 | 685 | |||
| GenBank mRNA | AK289369.1 | - | 9 | 768 | ||
| AK298680.1 | - | 8 | 648 | |||
| AK314466.1 | - | 10 | 809 | |||
| AL834477.1 | - | 9 | 744 | |||
| BC037386.1 | - | 7 | 643 | 4830820 | ||
| BC075801.1 | - | 9 | 744 | 5226828 | ||
| BC105789.1 | - | 10 | 813 | |||

PARP8
Genomic Map