siRNA Details
| Gene Symbol: ZMAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024645.2 | NP_078921.1 | 5, 6 | 731 | ||
| GenBank mRNA | AK023904.1 | - | 5, 6 | 731 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZMAT4| Gene Symbol: ZMAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024645.2 | NP_078921.1 | 5, 6 | 731 | ||
| GenBank mRNA | AK023904.1 | - | 5, 6 | 731 | ||