siRNA Details
| Gene Symbol: ANKRD55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024669.2 | NP_078945.2 | 10 | 1564 | ||
| XM_011543646.1 | XP_011541948.1 | 4 | 878 | |||
| XM_017009852.1 | XP_016865341.1 | 10 | 1476 | |||
| XM_017009853.1 | XP_016865342.1 | 9 | 1448 | |||
| XM_017009854.1 | XP_016865343.1 | 7 | 2798 | |||
| GenBank mRNA | AK021857.1 | - | 2 | 617 | ||
| AK123389.1 | - | 10 | 1564 | |||
| BC106914.2 | - | 2 | 784 | 40032109 | ||
| BC106915.2 | - | 2 | 784 | 40032110 | ||

ANKRD55
Genomic Map