siRNA Details
| Gene Symbol: CXorf36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024689.2 | NP_078965.2 | 3 | 1334 | ||
| GenBank mRNA | AK024165.1 | - | 3 | 1335 | ||
| BC035375.1 | - | 3 | 1270 | |||
| BC137528.1 | - | 3 | 1322 | |||
| BC137529.1 | - | 3 | 1322 | |||

CXorf36
Genomic Map