siRNA Details
| Gene Symbol: NIPAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321635.1 | NP_001308564.1 | 3 | 583 | ||
| NM_001321636.1 | NP_001308565.1 | 3 | 583 | |||
| NM_024759.2 | NP_079035.1 | 3 | 583 | |||
| NR_135745.1 | 3 | 583 | ||||
| NR_135746.1 | 3 | 583 | ||||
| NR_135747.1 | 3 | 583 | ||||
| XM_011517302.2 | XP_011515604.1 | 3 | 1617 | |||
| GenBank mRNA | AK024017.1 | - | 3 | 583 | ||
| AW082976.1 | - | 3 | 372 | |||
| BC132687.1 | - | 3 | 526 | 40146709 | ||
| BC144055.1 | - | 3 | 526 | 9052575 | ||
| BP346328.1 | - | 2 | 173 | |||
| DA184376.1 | - | 3 | 262 | |||

NIPAL2
Genomic Map