siRNA Details
| Gene Symbol: NIPAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321635.1 | NP_001308564.1 | 11 | 2153 | ||
| NM_001321636.1 | NP_001308565.1 | 10 | 2058 | |||
| NM_024759.2 | NP_079035.1 | 12 | 2181 | |||
| NR_135745.1 | 12 | 2317 | ||||
| NR_135746.1 | 13 | 2439 | ||||
| NR_135747.1 | 12 | 2377 | ||||
| GenBank mRNA | AK024017.1 | - | 12 | 2181 | ||

NIPAL2
Genomic Map