siRNA Details
| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 8 | 1630 | |||
| GenBank mRNA | AK022762.1 | - | 8 | 1567 | ||
| AK124860.1 | - | 1 | 871 | |||
| AK295771.1 | - | 9 | 1592 | |||
| AY359125.1 | - | 5 | 791 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF767P| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 8 | 1630 | |||
| GenBank mRNA | AK022762.1 | - | 8 | 1567 | ||
| AK124860.1 | - | 1 | 871 | |||
| AK295771.1 | - | 9 | 1592 | |||
| AY359125.1 | - | 5 | 791 | |||