siRNA Details
| Gene Symbol: POF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024921.3 | NP_079197.3 | 17 | 2951 | ||
| XM_005262203.3 | XP_005262260.1 | 17 | 2864 | |||
| GenBank mRNA | AF309774.1 | - | 17 | 2952 | ||
| AK025039.1 | - | 13 | 2542 | |||
| AK025080.1 | - | 9 | 1916 | |||
| AK026445.1 | - | 5 | 1443 | |||
| AK225193.1 | - | 9 | 1916 | |||

POF1B
Genomic Map