siRNA Details
| Gene Symbol: ABHD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319305.1 | NP_001306234.1 | 11 | 1181 | ||
| NM_001319306.1 | NP_001306235.1 | 10 | 1136 | |||
| NM_001319307.1 | NP_001306236.1 | 11 | 1199 | |||
| XM_005263238.4 | XP_005263295.1 | 11 | 1232 | |||
| XM_006714318.3 | XP_006714381.1 | 12 | 1277 | |||
| XM_006714320.3 | XP_006714383.1 | 12 | 1065 | |||
| XM_006714322.3 | XP_006714385.1 | 8 | 615 | |||
| XM_011532272.2 | XP_011530574.1 | 13 | 1344 | |||
| XM_011532273.2 | XP_011530575.1 | 12 | 1299 | |||
| XM_011532274.2 | XP_011530576.1 | 11 | 1175 | |||
| XM_011532276.1 | XP_011530578.1 | 11 | 1175 | |||
| XM_011532278.2 | XP_011530580.1 | 10 | 1038 | |||
| XM_011532279.2 | XP_011530581.1 | 5 | 551 | |||
| XM_017008631.1 | XP_016864120.1 | 10 | 1130 | |||
| XM_017008632.1 | XP_016864121.1 | 5 | 465 | |||
| GenBank mRNA | AK024759.1 | - | 10 | 948 | ||
| BC034253.1 | - | 10 | 1014 | 4339231 | ||
| BC111400.1 | - | 10 | 1116 | |||
| BC128142.1 | - | 10 | 886 | 40112521 | ||
| BC128143.1 | - | 10 | 829 | 40112522 | ||

ABHD18
Genomic Map