siRNA Details
| Gene Symbol: CXorf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025159.2 | NP_079435.1 | 3 | 1089 | ||
| GenBank mRNA | AK021639.1 | - | 3 | 970 | ||
| BC020611.1 | - | 3 | 1017 | 4768735 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXorf21| Gene Symbol: CXorf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025159.2 | NP_079435.1 | 3 | 1089 | ||
| GenBank mRNA | AK021639.1 | - | 3 | 970 | ||
| BC020611.1 | - | 3 | 1017 | 4768735 | ||