siRNA Details
| Gene Symbol: THBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003247.3 | NP_003238.2 | 7 | 1272 | ||
| GenBank mRNA | AK292429.1 | - | 7 | 1229 | ||
| BC146676.1 | - | 6 | 1079 | 40148868 | ||
| BC150175.1 | - | 6 | 1083 | 40147958 | ||
| L12350.1 | - | 7 | 1191 | |||

THBS2
Genomic Map