siRNA Details
| Gene Symbol: AKNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317950.1 | NP_001304879.1 | 3 | 795 | ||
| NM_001317952.1 | NP_001304881.1 | 2 | 353 | |||
| NM_030767.5 | NP_110394.3 | 3 | 740 | |||
| XM_005252244.2 | XP_005252301.1 | 5 | 912 | |||
| XM_005252245.1 | XP_005252302.1 | 4 | 770 | |||
| XM_005252247.4 | XP_005252304.1 | 3 | 791 | |||
| XM_005252248.2 | XP_005252305.1 | 3 | 908 | |||
| XM_006717294.1 | XP_006717357.1 | 3 | 724 | |||
| XM_006717295.2 | XP_006717358.1 | 3 | 908 | |||
| XM_011519063.2 | XP_011517365.1 | 2 | 403 | |||
| XM_011519064.2 | XP_011517366.1 | 3 | 908 | |||
| XM_011519065.2 | XP_011517367.2 | 3 | 383 | |||
| XM_011519066.2 | XP_011517368.1 | 3 | 908 | |||
| XM_017015172.1 | XP_016870661.1 | 4 | 510 | |||
| GenBank mRNA | AB075848.1 | - | 3 | 913 | ||
| AK126136.1 | - | 3 | 724 | |||
| AK131082.2 | - | 3 | 795 | |||
| AK160382.1 | - | 3 | 795 | |||
| AY703039.1 | - | 4 | 395 | |||
| AY703040.1 | - | 2 | 353 | |||
| AY703043.1 | - | 2 | 713 | |||
| AY703044.1 | - | 1 | 407 | |||
| BC055285.1 | - | 3 | 740 | 6143155 | ||

AKNA
Genomic Map