siRNA Details
| Gene Symbol: AKNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317950.1 | NP_001304879.1 | 12 | 2874 | ||
| NM_001317952.1 | NP_001304881.1 | 11 | 2432 | |||
| NM_030767.5 | NP_110394.3 | 12 | 2819 | |||
| XM_005252244.2 | XP_005252301.1 | 14 | 2991 | |||
| XM_005252245.1 | XP_005252302.1 | 13 | 2849 | |||
| XM_005252247.4 | XP_005252304.1 | 12 | 2870 | |||
| XM_005252248.2 | XP_005252305.1 | 12 | 2987 | |||
| XM_006717294.1 | XP_006717357.1 | 12 | 2803 | |||
| XM_011519063.2 | XP_011517365.1 | 11 | 2482 | |||
| XM_011519064.2 | XP_011517366.1 | 12 | 2987 | |||
| XM_011519065.2 | XP_011517367.2 | 12 | 2462 | |||
| XM_017015172.1 | XP_016870661.1 | 13 | 2589 | |||
| GenBank mRNA | AF286341.1 | - | 2 | 374 | ||
| AK024431.1 | - | 10 | 1767 | |||
| AK074040.1 | - | 11 | 1862 | |||
| AK131082.2 | - | 12 | 2874 | |||
| AK160382.1 | - | 12 | 2875 | |||
| AL833220.1 | - | 9 | 1296 | |||
| AY703039.1 | - | 13 | 2474 | |||
| AY703040.1 | - | 11 | 2432 | |||
| AY703043.1 | - | 11 | 2792 | |||
| AY703044.1 | - | 10 | 2486 | |||
| AY703045.1 | - | 3 | 569 | |||
| BC055285.1 | - | 12 | 2819 | 6143155 | ||

AKNA
Genomic Map