siRNA Details
| Gene Symbol: THBS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003248.5 | NP_003239.2 | 1, 2 | 334 | ||
| XM_017009798.1 | XP_016865287.1 | 1, 2 | 314 | |||
| XM_017009799.1 | XP_016865288.1 | 1, 2 | 314 | |||
| XM_017009800.1 | XP_016865289.1 | 1, 2 | 314 | |||
| GenBank mRNA | AK313587.1 | - | 1, 2 | 150 | ||
| BC050456.1 | - | 2, 3 | 418 | 6191808 | ||
| Z19585.1 | - | 1, 2 | 122 | |||

THBS4
Genomic Map