siRNA Details
| Gene Symbol: TRAPPC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031466.7 | NP_113654.4 | 1, 2 | 302 | ||
| XM_011517326.2 | XP_011515628.1 | 1, 2 | 302 | |||
| XM_011517328.2 | XP_011515630.1 | 1, 2 | 302 | |||
| XM_017013893.1 | XP_016869382.1 | 1, 2 | 302 | |||
| GenBank mRNA | AK128755.1 | - | 1, 2 | 303 | ||

TRAPPC9
Genomic Map