siRNA Details
| Gene Symbol: ARMC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161009.2 | NP_001154481.1 | 6 | 1300 | ||
| NM_001161010.2 | NP_001154482.1 | 6 | 1228 | |||
| NM_001161011.2 | NP_001154483.1 | 5 | 1228 | |||
| NM_001161012.2 | NP_001154484.1 | 5 | 1123 | |||
| NM_001161013.2 | NP_001154485.1 | 4 | 1051 | |||
| NM_031905.4 | NP_114111.2 | 7 | 1405 | |||
| XM_011516601.2 | XP_011514903.1 | 6 | 1478 | |||
| XM_011516602.2 | XP_011514904.1 | 6 | 839 | |||
| XM_017012684.1 | XP_016868173.1 | 8 | 1586 | |||
| XM_017012685.1 | XP_016868174.1 | 7 | 1514 | |||
| XM_017012686.1 | XP_016868175.1 | 7 | 1409 | |||
| XM_017012687.1 | XP_016868176.1 | 6 | 818 | |||
| GenBank mRNA | AK075500.1 | - | 7 | 1146 | ||
| AK095169.1 | - | 2 | 1858 | |||
| AK225082.1 | - | 6 | 1033 | |||
| AK291818.1 | - | 6 | 1028 | |||
| AK301565.1 | - | 5 | 961 | |||
| AY150851.1 | - | 5 | 1123 | |||
| AY150852.1 | - | 6 | 1228 | |||
| AY150853.1 | - | 6 | 1300 | |||
| AY150854.1 | - | 7 | 1405 | |||
| BC003586.1 | - | 6 | 1020 | 3357276 | ||
| BQ440425.1 | - | 1 | 82 | |||

ARMC10
Genomic Map