siRNA Details
| Gene Symbol: FAM188B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032222.2 | NP_115598.2 | 4 | 687 | ||
| GenBank mRNA | AF538962.1 | - | 4 | 680 | ||
| AK026027.1 | - | 4 | 687 | |||
| AK128558.1 | - | 2 | 2871 | |||
| BC098569.1 | - | 4 | 634 | 30924492 | ||
| Gene Symbol: INMT-FAM188B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037598.1 | 6 | 1139 | |||
| GenBank mRNA | AK308762.1 | - | 6 | 1137 | ||

FAM188B
Genomic Map