siRNA Details
| Gene Symbol: FAM126A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032581.3 | NP_115970.2 | 11 | 1602 | ||
| XM_005249894.3 | XP_005249951.1 | 12 | 1942 | |||
| XM_005249895.2 | XP_005249952.1 | 12 | 1861 | |||
| XM_011515589.2 | XP_011513891.1 | 11 | 1453 | |||
| GenBank mRNA | AB030241.1 | - | 11 | 1508 | ||
| AK054887.1 | - | 2 | 385 | |||
| AK056319.1 | - | 11 | 1539 | |||
| AL833296.1 | - | 11 | 1766 | |||
| BC018710.1 | - | 11 | 1580 | 4649785 | ||
| BX640757.1 | - | 1 | 1900 | |||
| KU178808.1 | - | 10 | 1347 | |||
| KU178809.1 | - | 11 | 1643 | |||
| KU178810.1 | - | 11 | 1562 | |||

FAM126A
Genomic Map