siRNA Details
| Gene Symbol: FAM71F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282788.1 | NP_001269717.1 | 7 | 1523 | ||
| NM_001282789.1 | NP_001269718.1 | 8 | 1470 | |||
| NM_032599.3 | NP_115988.1 | 7 | 1529 | |||
| NR_104242.1 | 8 | 1827 | ||||
| NR_104243.1 | 9 | 1716 | ||||
| GenBank mRNA | AF367470.1 | - | 7 | 1482 | ||
| AK093161.1 | - | 8 | 1467 | |||
| BC037248.1 | - | 7 | 1462 | 5165912 | ||

FAM71F1
Genomic Map