siRNA Details
| Gene Symbol: FAM220A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037163.1 | NP_001032240.1 | 1, 2 | 395 | ||
| GenBank mRNA | AF533707.1 | - | 1, 2 | 390 | ||
| AK093156.1 | - | 1, 2 | 26 | |||
| AK311878.1 | - | 1, 2 | 360 | |||
| BC006110.1 | - | 1, 2 | 395 | 3139887 | ||
| BC140712.1 | - | 1, 2 | 39 | 9021580 | ||
| BC142977.1 | - | 1, 2 | 411 | 8860408 | ||
| BP354450.1 | - | 1, 2 | 409 | |||

FAM220A
Genomic Map