siRNA Details
| Gene Symbol: FAM220A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037163.1 | NP_001032240.1 | 2 | 1737 | ||
| GenBank mRNA | AF533707.1 | - | 2 | 1732 | ||
| AK093156.1 | - | 2 | 1367 | |||
| BC006110.1 | - | 2 | 1737 | 3139887 | ||
| BC142977.1 | - | 2 | 1753 | 8860408 | ||

FAM220A
Genomic Map