siRNA Details
| Gene Symbol: TMEM209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032842.3 | NP_116231.2 | 10, 11 | 1369 | ||
| XM_005250655.1 | XP_005250712.1 | 10, 11 | 1659 | |||
| XM_011516646.2 | XP_011514948.1 | 10, 11 | 1509 | |||
| GenBank mRNA | AK027709.1 | - | 10, 11 | 1279 | ||
| AK074732.1 | - | 10, 11 | 1302 | |||
| BC045618.1 | - | 10, 11 | 1277 | 5294911 | ||

TMEM209
Genomic Map