siRNA Details
| Gene Symbol: TSLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033035.4 | NP_149024.1 | 4 | 886 | ||
| NM_138551.4 | NP_612561.2 | 2 | 668 | |||
| NR_045089.1 | 5 | 2091 | ||||
| XM_011543698.1 | XP_011542000.1 | 3 | 490 | |||
| GenBank mRNA | AL833421.1 | - | 2 | 648 | ||
| BC016720.1 | - | 2 | 601 | |||
| BC022894.1 | - | 2 | 640 | |||
| BC040592.1 | - | 5 | 2092 | 5269764 | ||

TSLP
Genomic Map