siRNA Details
| Gene Symbol: C5orf30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316968.1 | NP_001303897.1 | 3 | 1192 | ||
| NM_001316969.1 | NP_001303898.1 | 3 | 908 | |||
| NM_033211.3 | NP_149988.1 | 3 | 952 | |||
| GenBank mRNA | AF038182.1 | - | 3 | 846 | ||
| AK058020.1 | - | 3 | 888 | |||
| AK090558.1 | - | 3 | 1124 | |||
| BC009203.2 | - | 3 | 913 | 3628991 | ||

C5orf30
Genomic Map