siRNA Details
| Gene Symbol: FAM78A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033387.3 | NP_203745.2 | 2 | 1726 | ||
| XM_011518567.1 | XP_011516869.1 | 3 | 1269 | |||
| XM_011518568.2 | XP_011516870.1 | 2 | 1972 | |||
| GenBank mRNA | AK024434.1 | - | 3 | 2348 | ||
| AK095423.1 | - | 2 | 1754 | |||
| BC029924.2 | - | 2 | 1140 | |||
| BC037559.1 | - | 4 | 1447 | |||
| BC049388.2 | - | 2 | 1726 | 6050655 | ||

FAM78A
Genomic Map