siRNA Details
| Gene Symbol: TMEM203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053045.1 | NP_444273.1 | 1 | 1145 | ||
| GenBank mRNA | AF529372.1 | - | 1 | 186 | ||
| AX775751.1 | - | 1 | 1154 | |||
| BC009283.2 | - | 1 | 1145 | 4298098 | ||
| BC009461.1 | - | 1 | 1145 | 3538818 | ||
| BC067786.1 | - | 1 | 1157 | 5275460 | ||

TMEM203
Genomic Map