siRNA Details
| Gene Symbol: TSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058163.1 | NP_477511.1 | 3 | 231 | ||
| GenBank mRNA | BC007699.1 | - | 3 | 231 | ||
| BC011825.1 | - | 3 | 231 | 3830864 | ||
| U92980.1 | - | 2 | 135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSR2| Gene Symbol: TSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058163.1 | NP_477511.1 | 3 | 231 | ||
| GenBank mRNA | BC007699.1 | - | 3 | 231 | ||
| BC011825.1 | - | 3 | 231 | 3830864 | ||
| U92980.1 | - | 2 | 135 | |||