siRNA Details
| Gene Symbol: SMIM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135674.1 | NP_001129146.1 | 4 | 1282 | ||
| NM_001135675.1 | NP_001129147.1 | 4 | 890 | |||
| NM_001135676.1 | NP_001129148.1 | 4 | 575 | |||
| NM_138436.3 | NP_612445.2 | 4 | 661 | |||
| XM_005273398.4 | XP_005273455.1 | 4 | 1196 | |||
| GenBank mRNA | BC013035.2 | - | 4 | 623 | 3535608 | |
| BI463786.1 | - | 4 | 580 | |||
| DB519016.1 | - | 1 | 266 | |||

SMIM19
Genomic Map