siRNA Details
| Gene Symbol: MAGEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138703.4 | NP_619648.1 | 1 | 2029 | ||
| GenBank mRNA | AK094624.1 | - | 1 | 2021 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGEE2| Gene Symbol: MAGEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138703.4 | NP_619648.1 | 1 | 2029 | ||
| GenBank mRNA | AK094624.1 | - | 1 | 2021 | ||