siRNA Details
| Gene Symbol: TTC23L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317949.1 | NP_001304878.1 | 4 | 322 | ||
| NM_144725.3 | NP_653326.3 | 4 | 387 | |||
| XM_011513984.2 | XP_011512286.1 | 4 | 324 | |||
| XM_011513985.2 | XP_011512287.1 | 4 | 324 | |||
| XM_011513987.2 | XP_011512289.1 | 4 | 324 | |||
| XM_017009119.1 | XP_016864608.1 | 4 | 568 | |||
| XM_017009120.1 | XP_016864609.1 | 4 | 387 | |||
| XM_017009121.1 | XP_016864610.1 | 4 | 432 | |||
| XM_017009122.1 | XP_016864611.1 | 4 | 503 | |||
| XM_017009123.1 | XP_016864612.1 | 4 | 324 | |||
| GenBank mRNA | AK058168.1 | - | 4 | 387 | ||
| AY498865.1 | - | 3 | 284 | |||
| BC057775.1 | - | 4 | 359 | 5295348 | ||
| JN799997.1 | - | 4 | 322 | |||

TTC23L
Genomic Map