siRNA Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144962.2 | NP_659399.2 | 7 | 665 | ||
| XM_011544414.2 | XP_011542716.1 | 7 | 720 | |||
| GenBank mRNA | AY037148.1 | - | 7 | 638 | ||
| AY359109.1 | - | 7 | 661 | |||
| AY730275.1 | - | 7 | 616 | |||
| BC020779.1 | - | 7 | 657 | 4700840 | ||
| FJ224332.1 | - | 7 | 657 | |||

PEBP4
Genomic Map