siRNA Details
| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277171.1 | NP_001264100.1 | 4 | 1562 | ||
| NM_001277173.1 | NP_001264102.1 | 4 | 1565 | |||
| NM_144979.4 | NP_659416.1 | 4 | 1562 | |||
| XM_006714119.2 | XP_006714182.1 | 4 | 1559 | |||
| XM_006714120.1 | XP_006714183.1 | 4 | 1559 | |||
| XM_011531692.1 | XP_011529994.1 | 4 | 1513 | |||
| XM_011531693.1 | XP_011529995.1 | 4 | 1565 | |||
| XM_011531694.1 | XP_011529996.1 | 4 | 1416 | |||
| XM_011531695.2 | XP_011529997.1 | 4 | 1461 | |||
| XM_011531696.1 | XP_011529998.1 | 4 | 1416 | |||
| XM_011531697.2 | XP_011529999.1 | 4 | 1559 | |||
| GenBank mRNA | AK125877.1 | - | 4 | 1565 | ||
| AK302720.1 | - | 4 | 1499 | |||
| BC028588.2 | - | 4 | 1486 | 4830259 | ||

RBM46
Genomic Map